siRNA Details
| Gene Symbol: Gm4792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033209.1 | 3 | 500 | |||
| GenBank mRNA | AK053374.1 | - | 3 | 454 | ||
| BC092352.1 | - | 3 | 500 | 5400830 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4792| Gene Symbol: Gm4792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033209.1 | 3 | 500 | |||
| GenBank mRNA | AK053374.1 | - | 3 | 454 | ||
| BC092352.1 | - | 3 | 500 | 5400830 | ||