siRNA Details
| Gene Symbol: Mir764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030433.1 | 1 | 12 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mir764| Gene Symbol: Mir764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030433.1 | 1 | 12 | |||