siRNA Details
| Gene Symbol: 1700086L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030733.1 | 5 | 764 | |||
| NR_030734.1 | 5 | 759 | ||||
| NR_030735.1 | 4 | 537 | ||||
| GenBank mRNA | AK005679.1 | - | 3 | 431 | ||
| AK007020.1 | - | 4 | 533 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700086L19Rik| Gene Symbol: 1700086L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030733.1 | 5 | 764 | |||
| NR_030734.1 | 5 | 759 | ||||
| NR_030735.1 | 4 | 537 | ||||
| GenBank mRNA | AK005679.1 | - | 3 | 431 | ||
| AK007020.1 | - | 4 | 533 | |||