siRNA Details
| Gene Symbol: 5031426D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027890.1 | 1 | 5990 | |||
| GenBank mRNA | AK028985.1 | - | 1 | 2755 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB007613.2 | - | 1 | 617 | Chr. 2 - 6922702 - 6923332 [-] on Build GRCm38 | |

5031426D15Rik
Genomic Map