siRNA Details
| Gene Symbol: 1700022A21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003953.1 | 2 | 143 | |||
| GenBank mRNA | AK006230.1 | - | 2 | 143 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706316.1 | - | 2 | 143 | Chr. 5 - 24645453 - 24648338 [+] on Build GRCm38 | |

1700022A21Rik
Genomic Map