siRNA Details
| Gene Symbol: 1700022A21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003953.1 | 2 | 695 | |||
| GenBank mRNA | AK006230.1 | - | 2 | 694 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700022A21Rik| Gene Symbol: 1700022A21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003953.1 | 2 | 695 | |||
| GenBank mRNA | AK006230.1 | - | 2 | 694 | ||