siRNA Details
| Gene Symbol: Gt(ROSA)26Sor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_027008.1 | 2 | 290 | |||
| NR_027009.1 | 2 | 184 | ||||
| NR_027010.1 | 2 | 184 | ||||
| GenBank mRNA | AK021028.1 | - | 2 | 131 | ||
| BY207362.1 | - | 2 | 185 | |||
| BY727583.1 | - | 2 | 129 | |||
| U83174.1 | - | LIT3416 Rosa26.1 |
2 | 243 | ||
| U83175.1 | - | LIT3417 Rosa26.2 |
2 | 223 | ||

Gt(ROSA)26Sor
Genomic Map