siRNA Details
Gene Symbol: Gt(ROSA)26Sor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027008.1 | 1 | 31 | |||
NR_027009.1 | 1 | 31 | ||||
NR_027010.1 | 1 | 31 | ||||
GenBank mRNA | BY207362.1 | - | 1 | 32 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gt(ROSA)26Sor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027008.1 | 1 | 31 | |||
NR_027009.1 | 1 | 31 | ||||
NR_027010.1 | 1 | 31 | ||||
GenBank mRNA | BY207362.1 | - | 1 | 32 |