siRNA Details
| Gene Symbol: Nkx6-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027857.1 | 3 | 1323 | |||
| GenBank mRNA | AK008173.1 | - | 3 | 1324 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF471361.1 | - | 1 | 183 | Chr. 7 - 139581131 - 139581385 [-] on Build GRCm38 | |

Nkx6-2
Genomic Map