siRNA Details
| Gene Symbol: Nkx6-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027857.1 | 3 | 1534 | |||
| GenBank mRNA | AK008173.1 | - | 3 | 1536 | ||
| BY245395.1 | - | 1 | 38 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx6-2| Gene Symbol: Nkx6-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027857.1 | 3 | 1534 | |||
| GenBank mRNA | AK008173.1 | - | 3 | 1536 | ||
| BY245395.1 | - | 1 | 38 | |||