siRNA Details
| Gene Symbol: A930005H10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015487.1 | 1 | 268 | |||
| NR_027894.1 | 1 | 197 | ||||
| NR_027895.1 | 1 | 258 | ||||
| GenBank mRNA | AK007373.1 | - | 1 | 92 | ||
| AK020812.1 | - | 1 | 269 | |||
| AK081943.1 | - | 1 | 236 | |||
| AK137339.1 | - | 1 | 236 | |||
| BF019418.1 | - | 1 | 197 | |||

A930005H10Rik
Genomic Map