siRNA Details
| Gene Symbol: A930003A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015488.1 | 2 | 919 | |||
| NR_027896.1 | 3 | 813 | ||||
| GenBank mRNA | AK020802.1 | - | 2 | 543 | ||
| AK020919.1 | - | 2 | 573 | |||
| AK086168.1 | - | 3 | 814 | |||
| AK153879.1 | - | 2 | 921 | |||
| BC053981.1 | - | 2 | 865 | |||

A930003A15Rik
Genomic Map