siRNA Details
| Gene Symbol: A930003A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015488.1 | 2 | 1016 | |||
| NR_027896.1 | 3 | 910 | ||||
| GenBank mRNA | AK020802.1 | - | 2 | 640 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930003A15Rik| Gene Symbol: A930003A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015488.1 | 2 | 1016 | |||
| NR_027896.1 | 3 | 910 | ||||
| GenBank mRNA | AK020802.1 | - | 2 | 640 | ||