siRNA Details
| Gene Symbol: A930003A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015488.1 | 2 | 995 | |||
| NR_027896.1 | 3 | 889 | ||||
| GenBank mRNA | AK020802.1 | - | 2 | 619 | ||
| AK086168.1 | - | 3 | 890 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930003A15Rik| Gene Symbol: A930003A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015488.1 | 2 | 995 | |||
| NR_027896.1 | 3 | 889 | ||||
| GenBank mRNA | AK020802.1 | - | 2 | 619 | ||
| AK086168.1 | - | 3 | 890 | |||