siRNA Details
| Gene Symbol: A930003A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015488.1 | 2 | 892 | |||
| NR_027896.1 | 3 | 786 | ||||
| GenBank mRNA | AK020802.1 | - | 2 | 516 | ||
| AK020919.1 | - | 2 | 546 | |||
| AK086168.1 | - | 3 | 787 | |||
| AK153879.1 | - | 2 | 894 | |||
| BC053981.1 | - | 2 | 838 | |||

A930003A15Rik
Genomic Map