siRNA Details
| Gene Symbol: 2900041M22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015489.2 | 2 | 206 | |||
| GenBank mRNA | AK019332.1 | - | 2 | 79 | ||
| AK038849.1 | - | 2 | 110 | |||
| AK038850.1 | - | 2 | 110 | |||
| AK046435.1 | - | 2 | 207 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2900041M22Rik| Gene Symbol: 2900041M22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015489.2 | 2 | 206 | |||
| GenBank mRNA | AK019332.1 | - | 2 | 79 | ||
| AK038849.1 | - | 2 | 110 | |||
| AK038850.1 | - | 2 | 110 | |||
| AK046435.1 | - | 2 | 207 | |||