siRNA Details
| Gene Symbol: Gm4371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028311.1 | 2 | 1331 | |||
| GenBank mRNA | BB205215.2 | - | 1 | 366 | ||
| BY404711.1 | - | 1 | 95 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4371| Gene Symbol: Gm4371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028311.1 | 2 | 1331 | |||
| GenBank mRNA | BB205215.2 | - | 1 | 366 | ||
| BY404711.1 | - | 1 | 95 | |||