siRNA Details
| Gene Symbol: D4Ertd617e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029469.1 | 2 | 663 | |||
| GenBank mRNA | AK021029.1 | - | 2 | 663 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D4Ertd617e| Gene Symbol: D4Ertd617e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029469.1 | 2 | 663 | |||
| GenBank mRNA | AK021029.1 | - | 2 | 663 | ||