siRNA Details
| Gene Symbol: AI427809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033139.1 | 2 | 2006 | |||
| NR_033140.1 | 2 | 2096 | ||||
| GenBank mRNA | AK144542.1 | - | 2 | 2008 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI427809| Gene Symbol: AI427809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033139.1 | 2 | 2006 | |||
| NR_033140.1 | 2 | 2096 | ||||
| GenBank mRNA | AK144542.1 | - | 2 | 2008 | ||