siRNA Details
| Gene Symbol: AI427809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033139.1 | 2 | 1056 | |||
| NR_033140.1 | 2 | 1146 | ||||
| GenBank mRNA | AK144542.1 | - | 2 | 1058 | ||
| AK171862.1 | - | 2 | 1075 | |||
| BC147516.1 | - | 1 | 215 | 9056508 | ||
| BC147547.1 | - | 1 | 215 | 9056538 | ||

AI427809
Genomic Map