siRNA Details
| Gene Symbol: Gorasp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027343.1 | 2 | 285 | |||
| GenBank mRNA | AK002515.1 | - | 2 | 167 | ||
| AK168928.1 | - | 2 | 218 | |||
| AK210678.1 | - | 1 | 43 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gorasp2| Gene Symbol: Gorasp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027343.1 | 2 | 285 | |||
| GenBank mRNA | AK002515.1 | - | 2 | 167 | ||
| AK168928.1 | - | 2 | 218 | |||
| AK210678.1 | - | 1 | 43 | |||