siRNA Details
| Gene Symbol: C030034L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015499.2 | 2 | 2540 | |||
| GenBank mRNA | AK030949.1 | - | 2 | 2543 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C030034L19Rik| Gene Symbol: C030034L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015499.2 | 2 | 2540 | |||
| GenBank mRNA | AK030949.1 | - | 2 | 2543 | ||