siRNA Details
Gene Symbol: C030034L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015499.2 | 2 | 454 | |||
GenBank mRNA | AK030949.1 | - | 2 | 457 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C030034L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015499.2 | 2 | 454 | |||
GenBank mRNA | AK030949.1 | - | 2 | 457 |