siRNA Details
Gene Symbol: C030034L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015499.2 | 2 | 393 | |||
GenBank mRNA | AK030949.1 | - | 2 | 396 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB628125.1 | - | 3 | 508 | Chr. 3 - 9403075 - 9411287 [+] on Build GRCm38 |
