siRNA Details
Gene Symbol: A330040F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015503.1 | 2 | 706 | |||
GenBank mRNA | AK039403.1 | - | 660 | |||
AK052621.1 | - | 2 | 706 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A330040F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015503.1 | 2 | 706 | |||
GenBank mRNA | AK039403.1 | - | 660 | |||
AK052621.1 | - | 2 | 706 |