siRNA Details
| Gene Symbol: 5330426P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028300.1 | 2 | 1097 | |||
| GenBank mRNA | AK035047.1 | - | 1053 | |||
| AK045752.1 | - | 2 | 1062 | |||
| AK137010.1 | - | 2 | 1548 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5330426P16Rik| Gene Symbol: 5330426P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028300.1 | 2 | 1097 | |||
| GenBank mRNA | AK035047.1 | - | 1053 | |||
| AK045752.1 | - | 2 | 1062 | |||
| AK137010.1 | - | 2 | 1548 | |||