siRNA Details
| Gene Symbol: Firre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032694.1 | - | 1 | 359 | ||
| BC055934.1 | - | 3 | 888 | |||
| BC066100.1 | - | 15 | 1921 | |||
| KJ638680.1 | - | 4 | 317 | |||
| KJ638681.1 | - | 2 | 768 | |||
| KJ638682.1 | - | 3 | 313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Firre| Gene Symbol: Firre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032694.1 | - | 1 | 359 | ||
| BC055934.1 | - | 3 | 888 | |||
| BC066100.1 | - | 15 | 1921 | |||
| KJ638680.1 | - | 4 | 317 | |||
| KJ638681.1 | - | 2 | 768 | |||
| KJ638682.1 | - | 3 | 313 | |||