siRNA Details
| Gene Symbol: Firre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC066100.1 | - | 13 | 1618 | ||
| KJ638680.1 | - | 1 | 14 | |||
| KJ638681.1 | - | 1 | 14 | |||
| KJ638682.1 | - | 1 | 14 | |||
| KJ638683.1 | - | 1 | 14 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Firre| Gene Symbol: Firre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC066100.1 | - | 13 | 1618 | ||
| KJ638680.1 | - | 1 | 14 | |||
| KJ638681.1 | - | 1 | 14 | |||
| KJ638682.1 | - | 1 | 14 | |||
| KJ638683.1 | - | 1 | 14 | |||