siRNA Details
| Gene Symbol: G630025P09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027913.1 | 1 | 1425 | |||
| GenBank mRNA | AK014547.1 | - | 1191 | |||
| AK036110.1 | - | 1063 | ||||
| AK137322.1 | - | 1 | 1426 | |||
| AK139243.1 | - | 1 | 1416 | |||
| AK165723.1 | - | 1 | 1426 | |||

G630025P09Rik
Genomic Map