siRNA Details
Gene Symbol: G630025P09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027913.1 | 1 | 2359 | |||
GenBank mRNA | AK014547.1 | - | 2125 | |||
AK036110.1 | - | 1998 | ||||
AK165723.1 | - | 1 | 2360 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: G630025P09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027913.1 | 1 | 2359 | |||
GenBank mRNA | AK014547.1 | - | 2125 | |||
AK036110.1 | - | 1998 | ||||
AK165723.1 | - | 1 | 2360 |