siRNA Details
| Gene Symbol: G630025P09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027913.1 | 1 | 2159 | |||
| GenBank mRNA | AK014547.1 | - | 1925 | |||
| AK036110.1 | - | 1798 | ||||
| AK165723.1 | - | 1 | 2160 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G630025P09Rik| Gene Symbol: G630025P09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027913.1 | 1 | 2159 | |||
| GenBank mRNA | AK014547.1 | - | 1925 | |||
| AK036110.1 | - | 1798 | ||||
| AK165723.1 | - | 1 | 2160 | |||