siRNA Details
| Gene Symbol: 2900060B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027901.1 | 1 | 355 | |||
| GenBank mRNA | AK050598.1 | - | 1 | 355 | ||
| AK083371.1 | - | 1 | 355 | |||
| AK169832.1 | - | 1 | 356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2900060B14Rik| Gene Symbol: 2900060B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027901.1 | 1 | 355 | |||
| GenBank mRNA | AK050598.1 | - | 1 | 355 | ||
| AK083371.1 | - | 1 | 355 | |||
| AK169832.1 | - | 1 | 356 | |||