siRNA Details
| Gene Symbol: Ppp2r3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028110.2 | 1163 | ||||
| NR_102716.1 | 1281 | |||||
| GenBank mRNA | AK054122.1 | - | 1163 | |||
| AK143212.1 | - | 1067 | ||||
| BC096544.1 | - | 470 | 6834351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp2r3d| Gene Symbol: Ppp2r3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028110.2 | 1163 | ||||
| NR_102716.1 | 1281 | |||||
| GenBank mRNA | AK054122.1 | - | 1163 | |||
| AK143212.1 | - | 1067 | ||||
| BC096544.1 | - | 470 | 6834351 | |||