siRNA Details
| Gene Symbol: Gm9079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004052.1 | 1 | 702 | |||
| GenBank mRNA | AV025664.2 | - | 1 | 457 | ||
| BC038498.1 | - | 1 | 724 | |||
| Gene Symbol: Tmem5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043098.1 | - | 3494 | |||

Gm9079
Genomic Map