siRNA Details
Gene Symbol: Gm14379 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026741.1 | 2 | 876 | |||
GenBank mRNA | AK040575.1 | - | 2 | 690 | ||
AK155183.1 | - | 2 | 877 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm14379 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026741.1 | 2 | 876 | |||
GenBank mRNA | AK040575.1 | - | 2 | 690 | ||
AK155183.1 | - | 2 | 877 |