siRNA Details
| Gene Symbol: Gm14379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026741.1 | 2 | 1355 | |||
| GenBank mRNA | AK040575.1 | - | 2 | 1169 | ||
| AK155183.1 | - | 2 | 1358 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14379| Gene Symbol: Gm14379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026741.1 | 2 | 1355 | |||
| GenBank mRNA | AK040575.1 | - | 2 | 1169 | ||
| AK155183.1 | - | 2 | 1358 | |||