siRNA Details
| Gene Symbol: Gm14379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026741.1 | 1, 2 | 367 | |||
| GenBank mRNA | AK040575.1 | - | 1, 2 | 181 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB636260.1 | - | 1, 2 | 178 | Chr. X - 7376114 - 7377014 [+] on Build GRCm38 | |

Gm14379
Genomic Map