siRNA Details
| Gene Symbol: Gm14379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026741.1 | 1 | 191 | |||
| GenBank mRNA | AK155183.1 | - | 1 | 192 | ||
| BY203807.1 | - | 1 | 192 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14379| Gene Symbol: Gm14379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026741.1 | 1 | 191 | |||
| GenBank mRNA | AK155183.1 | - | 1 | 192 | ||
| BY203807.1 | - | 1 | 192 | |||