siRNA Details
| Gene Symbol: Gm14379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026741.1 | 2 | 1831 | |||
| GenBank mRNA | AK155183.1 | - | 2 | 1834 | ||
| BY547625.1 | - | 1 | 342 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14379| Gene Symbol: Gm14379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026741.1 | 2 | 1831 | |||
| GenBank mRNA | AK155183.1 | - | 2 | 1834 | ||
| BY547625.1 | - | 1 | 342 | |||