siRNA Details
| Gene Symbol: BB031773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028391.1 | 4 | 2085 | |||
| GenBank mRNA | AK030945.1 | - | 4 | 2087 | ||
| AK040278.1 | - | 4 | 2084 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BB031773| Gene Symbol: BB031773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028391.1 | 4 | 2085 | |||
| GenBank mRNA | AK030945.1 | - | 4 | 2087 | ||
| AK040278.1 | - | 4 | 2084 | |||