siRNA Details
| Gene Symbol: Gm5088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002862.3 | 1 | 3113 | |||
| GenBank mRNA | AK048750.1 | - | 1 | 2561 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5088| Gene Symbol: Gm5088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002862.3 | 1 | 3113 | |||
| GenBank mRNA | AK048750.1 | - | 1 | 2561 | ||