siRNA Details
| Gene Symbol: 2310068J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028124.1 | 1 | 977 | |||
| GenBank mRNA | AK010113.1 | - | 1 | 979 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310068J16Rik| Gene Symbol: 2310068J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028124.1 | 1 | 977 | |||
| GenBank mRNA | AK010113.1 | - | 1 | 979 | ||