siRNA Details
| Gene Symbol: 9430060I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015525.1 | 2 | 1121 | |||
| GenBank mRNA | AK137082.1 | - | 520 | |||
| AK162459.1 | - | 520 | ||||
| AK170241.1 | - | 1 | 486 | |||
| BC090260.1 | - | 2 | 1121 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9430060I03Rik| Gene Symbol: 9430060I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015525.1 | 2 | 1121 | |||
| GenBank mRNA | AK137082.1 | - | 520 | |||
| AK162459.1 | - | 520 | ||||
| AK170241.1 | - | 1 | 486 | |||
| BC090260.1 | - | 2 | 1121 | |||