siRNA Details
| Gene Symbol: 0610012G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027897.1 | 1 | 1028 | |||
| GenBank mRNA | AK002600.1 | - | 1 | 998 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

0610012G03Rik| Gene Symbol: 0610012G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027897.1 | 1 | 1028 | |||
| GenBank mRNA | AK002600.1 | - | 1 | 998 | ||