siRNA Details
| Gene Symbol: 1700108J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015532.1 | 2 | 447 | |||
| GenBank mRNA | AK006443.1 | - | 2 | 417 | ||
| AK018942.1 | - | 2 | 448 | |||
| AK018955.1 | - | 2 | 418 | |||
| BY707218.1 | - | 2 | 448 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700108J01Rik| Gene Symbol: 1700108J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015532.1 | 2 | 447 | |||
| GenBank mRNA | AK006443.1 | - | 2 | 417 | ||
| AK018942.1 | - | 2 | 448 | |||
| AK018955.1 | - | 2 | 418 | |||
| BY707218.1 | - | 2 | 448 | |||