siRNA Details
| Gene Symbol: 1700108J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015532.1 | 1 | 34 | |||
| GenBank mRNA | AK018942.1 | - | 1 | 35 | ||
| BY707218.1 | - | 1 | 35 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700108J01Rik| Gene Symbol: 1700108J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015532.1 | 1 | 34 | |||
| GenBank mRNA | AK018942.1 | - | 1 | 35 | ||
| BY707218.1 | - | 1 | 35 | |||