siRNA Details
| Gene Symbol: 4933407K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029443.1 | 9 | 1582 | |||
| GenBank mRNA | AK030939.1 | - | 10 | 1583 | ||
| AK036260.1 | - | 1 | 1502 | |||
| AK053637.1 | - | 9 | 1696 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB617014.1 | - | 2 | 495 | Chr. X - 75727098 - 75746904 [-] on Build GRCm38 | |

4933407K13Rik
Genomic Map