siRNA Details
| Gene Symbol: 4933407K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029443.1 | 9 | 2131 | |||
| GenBank mRNA | AK016728.1 | - | 5 | 1213 | ||
| AK030939.1 | - | 10 | 2132 | |||
| AK036260.1 | - | 1 | 2052 | |||
| AK053637.1 | - | 9 | 2245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933407K13Rik| Gene Symbol: 4933407K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029443.1 | 9 | 2131 | |||
| GenBank mRNA | AK016728.1 | - | 5 | 1213 | ||
| AK030939.1 | - | 10 | 2132 | |||
| AK036260.1 | - | 1 | 2052 | |||
| AK053637.1 | - | 9 | 2245 | |||