siRNA Details
| Gene Symbol: 4933400F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015540.1 | 4 | 1266 | |||
| GenBank mRNA | AK016592.1 | - | 4 | 1268 | ||
| AK029704.1 | - | 4 | 1271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933400F21Rik| Gene Symbol: 4933400F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015540.1 | 4 | 1266 | |||
| GenBank mRNA | AK016592.1 | - | 4 | 1268 | ||
| AK029704.1 | - | 4 | 1271 | |||