siRNA Details
Gene Symbol: Gm11213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028583.1 | 2 | 452 | |||
NR_028584.1 | 2 | 494 | ||||
GenBank mRNA | AK131740.1 | - | 2 | 453 | ||
BC100490.1 | - | 494 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm11213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028583.1 | 2 | 452 | |||
NR_028584.1 | 2 | 494 | ||||
GenBank mRNA | AK131740.1 | - | 2 | 453 | ||
BC100490.1 | - | 494 |